Merck
  • Efficient genome replication of hepatitis B virus using adenovirus vector: a compact pregenomic RNA-expression unit.
Efficient genome replication of hepatitis B virus using adenovirus vector: a compact pregenomic RNA-expression unit.
Scientific reports (2017-02-06)
Mariko Suzuki, Saki Kondo, Manabu Yamasaki, Norie Matsuda, Akio Nomoto, Tetsuro Suzuki, Izumu Saito, Yumi Kanegae
ABSTRACT
The complicated replication mechanisms of hepatitis B virus (HBV) have impeded HBV studies and anti-HBV therapy development as well. Herein we report efficient genome replication of HBV applying adenovirus vectors (AdVs) showing high transduction efficiency. Even in primary hepatocytes derived from humanized mice the transduction efficiencies using AdVs were 450-fold higher compared than those using plasmids. By using an expression unit consisting of the CMV promoter, 1.03-copy HBV genome and foreign poly(A) signal, we successfully generated an improved AdV (HBV103-AdV) that efficiently provided 58 times more pregenomic RNA than previously reported AdVs. The HBV103-AdV-mediated HBV replication was easily and precisely detected using quantitative real-time PCR in primary hepatocytes as well as in HepG2 cells. Notably, when the AdV containing replication-defective HBV genome of 1.14 copy was transduced, we observed that HBV DNA-containing circular molecules (pseudo-ccc DNA) were produced, which were probably generated through homologous recombination. However, the replication-defective HBV103-AdV hardly yielded the pseudo-ccc, probably because the repeated sequences are vey short. Additionally, the efficacies of entecavir and lamivudine were quantitatively evaluated using this system at only 4 days postinfection with HBV103-AdVs. Therefore, this system offers high production of HBV genome replication and thus could become used widely.
Sigma-Aldrich
Entecavir, ≥98% (HPLC)