Merck
  • Turn-On Colorimetric Platform for Dual Activity Detection of Acid and Alkaline Phosphatase in Human Whole Blood.
Turn-On Colorimetric Platform for Dual Activity Detection of Acid and Alkaline Phosphatase in Human Whole Blood.
Chemistry, an Asian journal (2016-09-13)
Qiong Hu, Baojing Zhou, Feng Li, Jinming Kong, Xueji Zhang
ABSTRACT
The activity detection of acid phosphatase (ACP) and alkaline phosphatase (ALP) is of great importance to the diagnosis and prognosis of related diseases. In this work, we report for the first time a turn-on colorimetric platform for the activity detection of ACP and ALP, by exploiting Cu(BCDS)
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)