Merck
  • Combination of AZD2281 (Olaparib) and GX15-070 (Obatoclax) results in synergistic antitumor activities in preclinical models of pancreatic cancer.
Combination of AZD2281 (Olaparib) and GX15-070 (Obatoclax) results in synergistic antitumor activities in preclinical models of pancreatic cancer.
Cancer letters (2014-02-19)
Shaohua Chen, Guan Wang, Xiaojia Niu, Jianyun Zhao, Wenxi Tan, Hebin Wang, Lijing Zhao, Yubin Ge
ABSTRACT
In this study, we explored the antitumor activities of the PARP inhibitor AZD2281 (Olaparib) and the pan-Bcl-2 inhibitor GX15-070 (Obatoclax) in six pancreatic cancer cell lines. While both agents were able to cause growth arrest and limited apoptosis, the combination of the two was able to synergistically cause growth arrest and non-apoptotic cell death. Furthermore, in an in vivo xenograft model, the combination caused substantially increased tumor necrosis compared to either treatment alone. Our results support further investigation of the combination of Bcl-2 and PARP inhibitors for the treatment of pancreatic cancer.