Merck
  • Transplantation of SIRT1-engineered aged mesenchymal stem cells improves cardiac function in a rat myocardial infarction model.
Transplantation of SIRT1-engineered aged mesenchymal stem cells improves cardiac function in a rat myocardial infarction model.
The Journal of heart and lung transplantation : the official publication of the International Society for Heart Transplantation (2014-07-19)
Xianbao Liu, Huiqiang Chen, Wei Zhu, Han Chen, Xinyang Hu, Zhi Jiang, Yinchuan Xu, Yu Zhou, Kan Wang, Lihan Wang, Panpan Chen, Hengxun Hu, Chen Wang, Na Zhang, Qunchao Ma, Mingyuan Huang, Dexing Hu, Ling Zhang, Rongrong Wu, Yaping Wang, Qiyuan Xu, Hong Yu, Jian'an Wang
ABSTRACT
Previous studies have demonstrated that biological aging has a negative influence on the therapeutic effects of mesenchymal stem cells (MSCs)-based therapy. Using a rat myocardial infarction (MI) model, we tested the hypothesis that silent mating type information regulation 2 homolog 1 (SIRT1) may ameliorate the phenotype and improve the function of aged MSCs and thus enhance the efficacy of aged MSCs-based therapy. Sixty female rats underwent left anterior descending coronary artery ligation and were randomly assigned to receiving: intramyocardial injection of cell culture medium (DMEM group); SIRT1 overexpression vector-treated aged MSCs (SIRT1-aged MSCs group) obtained from aged male SD rats or empty vector-treated aged MSCs (vector-aged MSCs group). Another 20 sham-operated rats that underwent open-chest surgery without coronary ligation or any other intervention served as controls. SIRT1-aged MSC group exhibited enhanced blood vessel density in the border zone of MI hearts, which was associated with reduced cardiac remodeling, leading to improved cardiac performance. Consistent with the in vivo data, our in vitro experiments also demonstrated that SIRT1 overexpression ameliorated aged MSCs senescent phenotype and recapitulated the pro-angiogenesis property of MSCs and conferred the anti-stress response capabilities, as indicated by increases in pro-angiogenic factors, angiopoietin 1 (Ang1) and basic fibroblast growth factor (bFGF), expressions and a decrease in anti-angiogenic factor thrombospondin-1 (TBS1) at mRNA levels, and increases in Bcl-2/Bax ratio at protein level. Up-regulating SIRT1 expression could enhance the efficacy of aged MSCs-based therapy for MI as it relates to the amelioration of senescent phenotype and hence improved biological function of aged MSCs.