- Regulation of catalytic activity of S6 kinase 2 during cell cycle.
Regulation of catalytic activity of S6 kinase 2 during cell cycle.
Molecular and cellular biochemistry (2007-09-06)
Derek Boyer, Rene Quintanilla, Kay K Lee-Fruman
PMID17786541
ABSTRACT
Ribosomal S6 kinase 2 (S6K2) is one of the kinases regulated by the mammalian target of rapamycin (mTOR) signaling pathway. Although it has been identified as a kinase homologous to S6K1, evidence suggests that the two kinases have non-overlapping functions, and the biological function of S6K2 still remains unknown. In order to identify the cell cycle stage(s) during which S6K2 plays a role, we assessed changes in the catalytic activity of S6K2 throughout the cell cycle. Our data show that S6K2 is active throughout the cell cycle with higher activity in G2 and M phases. We also show that S6K1 activity peaks sharply during M phase. Our data suggest that S6K1 and S6K2 likely play yet-unknown roles in G2 and M phases.