Merck
  • Synthesis of novel aza-analogues of tiazofurin with 2-[5,5-bis(hydroxymethyl)pyrrolidin-2-yl] framework as sugar mimic.
Synthesis of novel aza-analogues of tiazofurin with 2-[5,5-bis(hydroxymethyl)pyrrolidin-2-yl] framework as sugar mimic.
Nucleosides, nucleotides & nucleic acids (2012-01-20)
Ewa Mironiuk-Puchalska, Mariola Koszytkowska-Stawińska, Wojciech Sas, Erik De Clercq, Lieve Naesens
ABSTRACT
The novel aza-analogues of tiazofurin (TZF) with 2-[5,5-bis(hydroxymethyl)pyrrolidin-2-yl] moiety, as sugar mimic, were synthesized from O,O-cyclohexylidene derivative of 4,4-bis(hydroxymethyl)-4-nitrobutanal in multi-gram scale. The synthetic route consisted of three stages: (i) the synthesis of corresponding derivative of 5,5-bis(hydroxymethyl)pyrrolidine-2-carbonitrile, (ii) the construction of ethyl thiazole-2-carboxylate part by the conversion of the pyrrolidine-2-carbonitrile into the N-trifluoroacetyl derivative followed by cyclocondensation with L-cysteine ethyl ester and then by dehydrogenation, and (iii) the final transformation of the ethyl thiazole-4-carboxylate into the aza-analogues of TZF. The TZF aza-analogues were evaluated for their antiviral activities in cell-culture-based assays.
Sigma-Aldrich
L-Cysteine ethyl ester hydrochloride, 98%