Merck
  • Development of an amperometric biosensor based on peroxidases to quantify citrinin in rice samples.
Development of an amperometric biosensor based on peroxidases to quantify citrinin in rice samples.
Bioelectrochemistry (Amsterdam, Netherlands) (2013-02-19)
Vanesa Gimena Lourdes Zachetti, Adrian Marcelo Granero, Sebastián Noel Robledo, María Alicia Zon, Héctor Fernández
ABSTRACT
An amperometric biosensor based on horseradish peroxidase (EC1.11.1.7,H2O2-oxide-reductases) to determine the content of citrinin mycotoxin in rice samples is proposed by the first time. The method uses carbon paste electrodes filled up with multi-walled carbon nanotubes embedded in a mineral oil, horseradish peroxidase, and ferrocene as a redox mediator. The biosensor is covered externally with a dialysis membrane, which is fixed to the body side of the electrode with a Teflon laboratory film, and an O-ring. The reproducibility and the repeatability were of 7.0% and 3.0%, respectively, showing a very good biosensor performance. The calibration curve was linear in a concentration range from 1 to 11.6nM. The limits of detection and quantification were 0.25nM and 0.75nM, respectively. For comparison, the citrinin content in rice samples was also determined by fluorimetric measurements. A very good correlation was obtained between the electrochemical and spectrophotometric methods.
Product Description
Supelco
Citrinin solution, ~100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Citrinin, from Penicillium citrinum, ≥98% (HPLC)