Merck
  • Curcuminoid analogs inhibit nitric oxide production from LPS-activated microglial cells.
Curcuminoid analogs inhibit nitric oxide production from LPS-activated microglial cells.
Journal of natural medicines (2011-10-14)
Jiraporn Tocharus, Sataporn Jamsuwan, Chainarong Tocharus, Chatchawan Changtam, Apichart Suksamrarn
ABSTRACT
The chemically modified analogs, the demethylated analogs 4-6, the tetrahydro analogs 7-9 and the hexahydro analogs 10-12, of curcumin (1), demethoxycurcumin (2) and bisdemethoxycurcumin (3) were evaluated for their inhibitory activity on lipopolysaccharide activated nitric oxide (NO) production in HAPI microglial cells. Di-O-demethylcurcumin (5) and O-demethyldemethoxycurcumin (6) are the two most potent compounds that inhibited NO production. The analogs 5 and 6 were twofold and almost twofold more active than the parent curcuminoids 1 and 2, respectively. Moreover, the mRNA expression level of inducible NO synthase was inhibited by these two compounds. The strong neuroprotective activity of analogs 5 and 6 provide potential alternative compounds to be developed as therapeutics for neurological disorders associated with activated microglia.
Sigma-Aldrich
Demethoxycurcumin, ≥98% (HPLC)
Supelco
Demethoxycurcumin, analytical standard
USP
Desmethoxycurcumin, United States Pharmacopeia (USP) Reference Standard