Merck
  • Intracellular localization of the 55-kD actin-bundling protein in cultured cells: spatial relationships with actin, alpha-actinin, tropomyosin, and fimbrin.
Intracellular localization of the 55-kD actin-bundling protein in cultured cells: spatial relationships with actin, alpha-actinin, tropomyosin, and fimbrin.
The Journal of cell biology (1986-08-01)
S Yamashiro-Matsumura, F Matsumura
ABSTRACT
The 55-kD protein is a new actin-bundling protein purified from HeLa cells (Yamashiro-Matsumura, S., and F. Matsumura, 1985, J. Biol. Chem., 260:5087-5097). We have prepared monoclonal antibodies against the 55-kD protein and examined its intracellular localization, as well as its spatial relationships with other components of microfilaments in cultured cells by double-label immunofluorescence. The localization of the 55-kD protein is similar to that of actin. The antibody to the 55-kD protein stained strongly both microspikes and stress fibers. The 55-kD protein was found from the basal portions to the extremities of microspikes while alpha-actinin was localized only in the basal portions. In stress fibers, the 55-kD protein was found rather continuously in comparison to the periodic localizations of alpha-actinin and tropomyosin. Although fimbrin is located in microspikes and ruffling membranes, fimbrin is hardly found in stress fibers unlike the 55-kD protein. These observations coupled with the actin-bundling activity of the 55-kD protein imply that the 55-kD protein is involved in the formation of microfilament bundles in both microspikes and stress fibers.