Skip to Content
Merck
Tamoxifen solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Tamoxifen

Tamoxifen solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
(Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene, trans-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethylamine
Linear Formula:
C6H5C(C2H5)=C(C6H5)C6H4OCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
371.51

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Package Size
Special Grade
Technique