Skip to Content
Merck
Codeine solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Codeine solution

Codeine solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C18H21NO3
CAS Number:
Molecular Weight:
299.36
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique