Skip to Content
Merck
Menidazole VETRANAL®, analytical standard

Tinidazole Related Compound A

Menidazole VETRANAL®, analytical standard
Synonym(s):
2-Methyl-4(5)-nitroimidazole, 2-Methyl-5-nitro-1H-imidazole
Empirical Formula (Hill Notation):
C4H5N3O2
CAS Number:
Molecular Weight:
127.10
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique