![N,N-Dimethyl-2-[1-(methylsulfonyl)-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/329/938/f2fbd23e-6a9d-441a-a765-ec4ee32f7eda/640/f2fbd23e-6a9d-441a-a765-ec4ee32f7eda.png)
N,N-Dimethyl-2-[1-(methylsulfonyl)-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine
![N,N-Dimethyl-2-[1-(methylsulfonyl)-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/329/938/f2fbd23e-6a9d-441a-a765-ec4ee32f7eda/640/f2fbd23e-6a9d-441a-a765-ec4ee32f7eda.png)
Empirical Formula (Hill Notation):
C16H21N5O2S
Molecular Weight:
347.44
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1