Skip to Content
Merck
Erythrosine B (C.I. 45430) for microscopy Certistain®

Erythrosine B (C.I. 45430)

Erythrosine B (C.I. 45430) for microscopy Certistain®
Synonym(s):
Erythrosine B (C.I. 45430)
Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing