Skip to Content
Merck
DL-Lactic acid 88% for analysis EMPARTA®

DL-Lactic acid 88%

DL-Lactic acid 88% for analysis EMPARTA®
Synonym(s):
DL-Lactic acid, 2-Hydroxypropionic acid
Linear Formula:
CH3CH(OH)COOH
CAS Number:
Molecular Weight:
90.08

Products

Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing