Sodium azide
Products
Product Category
Brand
Biological Source
Color
Formula Weight
pH Value
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing