Skip to Content
Merck
Trazodone hydrochloride ≥99% (HPLC), powder

Trazodone hydrochloride

Trazodone hydrochloride ≥99% (HPLC), powder
Synonym(s):
2-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one hydrochloride
Empirical Formula (Hill Notation):
C19H22ClN5O · HCl
CAS Number:
Molecular Weight:
408.32
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing