Skip to Content
Merck
2-Butanone solution certified reference material, 2000 μg/mL in methanol: water (9:1)

2-Butanone solution

2-Butanone solution certified reference material, 2000 μg/mL in methanol: water (9:1)
Synonym(s):
2-Butanone, Ethyl methyl ketone, MEK, Methyl ethyl ketone
Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11

Products

Boiling Point (°C)
Formula Weight
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1