Skip to Content
Merck
Magnesium chloride hexahydrate tested according to Ph. Eur.

Magnesium chloride hexahydrate

Magnesium chloride hexahydrate tested according to Ph. Eur.
Synonym(s):
Magnesium chloride hexahydrate
Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Showing 1-20 of 24
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2