Skip to Content
HK
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zinc chloride
Synonym(s):
Dichlorozinc, Zinc chloride
Linear Formula:
ZnCl
2
CAS Number:
7646-85-7
Molecular Weight:
136.30
EC Number:
231-592-0
Products
Product Category
salts (10)
Brand
SAFC (2)
Sigma-Aldrich (11)
Supelco (2)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reagent Type
Core
Special Grade
Anhydrous (4)
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
208086
Zinc chloride,
reagent grade, ≥98%
Expand
View Pricing
793523
Zinc chloride,
anhydrous, free-flowing, Redi-Dri
™
, reagent grade, ≥98%
Expand
View Pricing
429430
Zinc chloride,
anhydrous, powder, ≥99.995% trace metals basis
Expand
View Pricing
1.08816
Zinc chloride,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
Z0152
Zinc chloride,
BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
229997
Zinc chloride,
99.999% trace metals basis
Expand
View Pricing
211273
Zinc chloride,
ACS reagent, ≥97%
Expand
View Pricing
746355
Zinc chloride,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥97%
Expand
View Pricing
31650
Zinc chloride,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Expand
View Pricing
1.28221
Zinc chloride,
EMPROVE
®
EXPERT, Ph. Eur., BP, USP
Expand
View Pricing
14422
Zinc chloride,
puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Expand
View Pricing
1.28220
Zinc chloride,
EMPROVE
®
API, Ph. Eur., BP, JP, USP
Expand
View Pricing
450111
Zinc chloride,
AnhydroBeads
™
, amorphous, −10 mesh, 99.99% trace metals basis
Expand
View Pricing
1.93623
Zinc chloride,
EMPLURA
®
Expand
View Pricing
456845
Zinc chloride,
AnhydroBeads
™
, amorphous, −10 mesh, 99.999% trace metals basis
Expand
View Pricing