Skip to Content
HK
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Casein from bovine milk
Synonym(s):
Casein from bovine milk, Casein, Bovine Milk, Carbohydrate and Fatty Acid Free, Casein, Bovine Milk
CAS Number:
9000-71-9
EC Number:
232-555-1
Products
Product Category
proteins (11)
substrates (1)
Brand
Sigma-Aldrich (17)
Supelco (1)
Biological Source
Melting Point (°C)
Physical Form
Product Line
Purity
Tissue
Quality Segment
Technique
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
C7078
Casein from bovine milk,
technical grade
Expand
View Pricing
C6905
Casein from bovine milk,
BioUltra, ≥98% (PAGE)
Expand
View Pricing
C5890
Casein from bovine milk,
suitable for substrate for protein kinase (after dephosphorylation), purified powder
Expand
View Pricing
C3400
Casein from bovine milk,
powder
Expand
View Pricing
C0406
Casein from bovine milk,
≥70% (PAGE), lyophilized powder
Expand
View Pricing
C6780
Casein from bovine milk,
≥70% α
s
-casein basis (electrophoresis), lyophilized powder
Expand
View Pricing
E0789
Casein from bovine milk,
Expand
View Pricing
C9801
Casein from bovine milk,
essentially salt-free, lyophilized powder
Expand
View Pricing
218682
Casein from bovine milk,
Casein, Bovine Milk, Carbohydrate and Fatty Acid Free, is a high molecular weight phosphoprotein. Useful as a substrate for various proteases. Supplied as Hammarsten grade material.
Expand
View Pricing
218680
Casein from bovine milk,
Casein, Bovine Milk, CAS 9000-71-9, is a high molecular weight phosphoprotein. Useful as a substrate for bromelain, chymotrypsin, pepsin, trypsin, and other proteases.
Expand
View Pricing
SRE0081
Casein from bovine milk,
Suitable for manufacturing of diagnostic kits and reagents, Casein, alkali treated
Expand
View Pricing
C6554
Casein from bovine milk,
BioReagent, suitable for insect cell culture
Expand
View Pricing
C5679
Casein from bovine milk,
BioReagent, suitable for insect cell culture
Expand
View Pricing
SRE0040
Casein from bovine milk,
Suitable for manufacturing of diagnostic kits and reagents
Expand
View Pricing
E3414
Casein from bovine milk,
powder
Expand
View Pricing
1.02244
Casein from bovine milk,
Expand
View Pricing
C8032
Casein from bovine milk,
lyophilized powder
Expand
View Pricing
C4032
Casein from bovine milk,
lyophilized powder
Expand
View Pricing
1.02241
Casein from bovine milk,
soluble in alkali, Technipur
®
Expand
View Pricing
C7906
Casein from bovine milk,
for use in the Total Dietary Fiber Control Kit, TDF-C10
Expand
View Pricing