Skip to Content
Merck
Charcoal wood powder

Carbon

Charcoal wood powder
Synonym(s):
Carbon, Charcoal wood
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Particle Size (µm)
Showing 1-3 of 3