US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity-5, NPD Detection
![US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity-5, NPD Detection suitable for GC](/deepweb/assets/sigmaaldrich/product/images/138/860/805679f1-007c-481b-ba96-5d2348136d7e/800/805679f1-007c-481b-ba96-5d2348136d7e.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-5, 30m × 0.25 mm I.D., 0.25 μm (28089-U)
oven
120 °C (3 min), 5 °C/min, to 300 °C (5 min)
inj. temp.
250 °C
detector
NPD, 320 °C
carrier gas
helium, 25 cm/sec @ 120 °C
injection
1.0 μL splitless (0.5 min)
liner
4 mm I.D., double taper
sample
40 ng on-column of a custom organophosphorus pesticides mix
Description
Legal Information
null