Merck
  • Isopentenoid synthesis in eukaryotic cells. An initiating role for post-translational control of 3-hydroxy-3-methylglutaryl coenzyme A reductase.
Isopentenoid synthesis in eukaryotic cells. An initiating role for post-translational control of 3-hydroxy-3-methylglutaryl coenzyme A reductase.
Archives of biochemistry and biophysics (1993-04-01)
M D Giron, C M Havel, J A Watson
ABSTRACT
Using insect (Drosophila) and rodent mammalian cell cultures we demonstrate that acute mevalonate-mediated suppression of 3-hydroxy-3-methylglutaryl coenzyme A was initiated in the absence of protein synthesis. In addition, insect and mammalian cells depleted (1 h) of putative regulatory post-isopentenyl-1-pyrophosphate metabolites by incubation with an inhibitor of 3-hydroxy-3-methylglutaryl coenzyme A synthase (L-659,699) accumulated 3-hydroxy-3-methylglutaryl coenzyme A reductase enzyme units. This accumulation resulted primarily from a decrease in the loss of 3-hydroxy-3-methylglutaryl coenzyme A reductase enzyme units rather than from an increase in enzyme synthesis. These observations suggest that the initial phase of mevalonate-mediated suppression of 3-hydroxy-3-methylglutaryl coenzyme A reductase activity was governed primarily by post-translational processes. An unknown rapidly turning over post-isopentenyl-1-pyrophosphate metabolite(s) is proposed as the agonist for these post-translational processes.
Sigma-Aldrich
Hymeglusin, ≥98% (HPLC), from Fusarium sp.