Merck
  • Addition of losartan to angiotensin-converting enzyme inhibitors improves insulin resistance in patients with chronic heart failure treated without β-blockers.
Addition of losartan to angiotensin-converting enzyme inhibitors improves insulin resistance in patients with chronic heart failure treated without β-blockers.
Circulation journal : official journal of the Japanese Circulation Society (2010-09-10)
Kazuhide Ogino, Masahiko Kato, Yoshiyuki Furuse, Yoshiharu Kinugasa, Yasuhiro Kaetsu, Einosuke Mizuta, Shinobu Sugihara, Katsunori Ishida, Kiyotaka Yanagihara, Ichiro Hisatome, Chiaki Shigemasa
ABSTRACT
Angiotensin II and insulin resistance (IR) have clinical implications in the pathophysiology of chronic heart failure (CHF). However, it is still unclear whether the combination of an angiotensin-receptor blocker and angiotensin-converting enzyme inhibitor (ACEI) improves IR in CHF patients who do not receive β-blockers. Thus, the aim of the present study was to evaluate the effects of losartan on glucose metabolism and inflammatory cytokines in CHF patients treated with ACEI but not β-blockers. The effect of losartan treatment for 16 weeks on IR was analyzed in 16 CHF patients in a randomized crossover trial. Insulin level and homeostasis model IR index (HOMA-IR) decreased significantly (P<0.05), but fasting plasma glucose did not change significantly. Serum tumor necrosis factor (TNF)-α, interleukin (IL)-6, and monocyte chemoattractant protein (MCP)-1 levels were significantly decreased with losartan (P<0.05). Furthermore, the changes in IL-6 and MCP-1 levels were significantly correlated with the reduction in HOMA-IR (P<0.05), but the change in TNF-α levels was not significantly correlated. The addition of losartan to ACEI therapy improved IR and decreased inflammatory cytokines in CHF patients who did not receive β-blockers.
Sigma-Aldrich
Imidapril hydrochloride, ≥98% (HPLC)