Merck
  • Generation of two gene corrected human isogenic iPSC lines (NCATS-CL6104 and NCATS-CL6105) from a patient line (NCATS-CL6103) carrying a homozygous p.R401X mutation in the NGLY1 gene using CRISPR/Cas9.
Generation of two gene corrected human isogenic iPSC lines (NCATS-CL6104 and NCATS-CL6105) from a patient line (NCATS-CL6103) carrying a homozygous p.R401X mutation in the NGLY1 gene using CRISPR/Cas9.
Stem cell research (2021-10-08)
Ivan Pavlinov, Atena Farkhondeh, Shu Yang, Miao Xu, Yu-Shan Cheng, Jeanette Beers, Jizhong Zou, Chengyu Liu, Matthew Might, Steven Rodems, Karsten Baumgärtel, Wei Zheng
ABSTRACT
NGLY1 deficiency is a rare recessive genetic disease caused by mutations in the NGLY1 gene which codes for N-glycanase 1 (NGLY1). Here, we report the generation of two gene corrected iPSC lines using a patient-derived iPSC line (NCATS-CL6103) that carried a homozygous p.R401X mutation in the NGLY1 gene. These lines contain either one (NCATS-CL6104) or two (NCATS-CL6105) CRISPR/Cas9 corrected alleles of NGLY1. This pair of NGLY1 mutation corrected iPSC lines can be used as a control for the NCATS-CL6103 which serves as a cell-based NGLY1 disease model for the study of the disease pathophysiology and evaluation of therapeutics under development.
Sigma-Aldrich
Milli-Mark® Anti-Nanog-Alexa Fluor 488 Antibody, NT, Milli-Mark®, from rabbit
Sigma-Aldrich
Anti-NGLY1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution