Merck
  • DNA spike-ins enable confident interpretation of SARS-CoV-2 genomic data from amplicon-based sequencing.
DNA spike-ins enable confident interpretation of SARS-CoV-2 genomic data from amplicon-based sequencing.
bioRxiv : the preprint server for biology (2021-03-25)
Kim A Lagerborg, Erica Normandin, Matthew R Bauer, Gordon Adams, Katherine Figueroa, Christine Loreth, Adrianne Gladden-Young, Bennett Shaw, Leah Pearlman, Erica S Shenoy, David Hooper, Virginia M Pierce, Kimon C Zachary, Daniel J Park, Bronwyn L MacInnis, Jacob E Lemieux, Pardis C Sabeti, Steven K Reilly, Katherine J Siddle
ABSTRACT
The rapid global spread and continued evolution of SARS-CoV-2 has highlighted an unprecedented need for viral genomic surveillance and clinical viral sequencing. Amplicon-based sequencing methods provide a sensitive, low-cost and rapid approach but suffer a high potential for contamination, which can undermine lab processes and results. This challenge will only increase with expanding global production of sequences by diverse research groups for epidemiological and clinical interpretation. We present an approach which uses synthetic DNA spike-ins (SDSIs) to track samples and detect inter-sample contamination through a sequencing workflow. Applying this approach to the ARTIC Consortium's amplicon design, we define a series of best practices for Illumina-based sequencing and provide a detailed characterization of approaches to increase sensitivity for low-viral load samples incorporating the SDSIs. We demonstrate the utility and efficiency of the SDSI method amidst a real-time investigation of a suspected hospital cluster of SARS-CoV-2 cases.
Sigma-Aldrich
KOD Hot Start Master Mix, Ready-to-use 2X mixture, containing KOD Hot Start DNA Polymerase, two monoclonal antibodies, ultrapure deoxynucleotides, and reaction buffer with MgSO4, optimized for convenient high fidelity PCR., ready-to-use solution, suitable for PCR