Merck
  • IL-7 is critical for homeostatic proliferation and survival of naive T cells.
IL-7 is critical for homeostatic proliferation and survival of naive T cells.
Proceedings of the National Academy of Sciences of the United States of America (2001-07-12)
J T Tan, E Dudl, E LeRoy, R Murray, J Sprent, K I Weinberg, C D Surh
ABSTRACT
In T cell-deficient conditions, naive T cells undergo spontaneous "homeostatic" proliferation in response to contact with self-MHC/peptide ligands. With the aid of an in vitro system, we show here that homeostatic proliferation is also cytokine-dependent. The cytokines IL-4, IL-7, and IL-15 enhanced homeostatic proliferation of naive T cells in vitro. Of these cytokines, only IL-7 was found to be critical; thus, naive T cells underwent homeostatic proliferation in IL-4(-) and IL-15(-) hosts but proliferated minimally in IL-7(-) hosts. In addition to homeostatic proliferation, the prolonged survival of naive T cells requires IL-7. Thus, naïve T cells disappeared gradually over a 1-month period upon adoptive transfer into IL-7(-) hosts. These findings indicate that naive T cells depend on IL-7 for survival and homeostatic proliferation.