Merck
  • Neutrophil degranulation interconnects over-represented biological processes in atrial fibrillation.
Neutrophil degranulation interconnects over-represented biological processes in atrial fibrillation.
Scientific reports (2021-02-05)
Makiri Kawasaki, Eva R Meulendijks, Nicoline W E van den Berg, Fransisca A Nariswari, Jolien Neefs, Robin Wesselink, Sarah W E Baalman, Aldo Jongejan, Tim Schelfhorst, Sander R Piersma, Thang V Pham, Wim J P van Boven, Antoine H G Driessen, Connie R Jimenez, Joris R de Groot
ABSTRACT
Despite our expanding knowledge about the mechanism underlying atrial fibrillation (AF), the interplay between the biological events underlying AF remains incompletely understood. This study aimed to identify the functionally enriched gene-sets in AF and capture their interconnection via pivotal factors, that may drive or be driven by AF. Global abundance of the proteins in the left atrium of AF patients compared to control patients (n = 3/group), and the functionally enriched biological processes in AF were determined by mass-spectrometry and gene set enrichment analysis, respectively. The data were validated in an independent cohort (n = 19-20/group). In AF, the gene-sets of innate immune system, metabolic process, cellular component disassembly and ion homeostasis were up-regulated, while the gene-set of ciliogenesis was down-regulated. The innate immune system was over-represented by neutrophil degranulation, the components of which were extensively shared by other gene-sets altered in AF. In the independent cohort, an activated form of neutrophils was more present in the left atrium of AF patients with the increased gene expression of neutrophil granules. MYH10, required for ciliogenesis, was decreased in the atrial fibroblasts of AF patients. We report the increased neutrophil degranulation appears to play a pivotal role, and affects multiple biological processes altered in AF.
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Anti-Calnexin, C-Terminal (575-593) Rabbit pAb, liquid, Calbiochem®