Merck
  • The phenotype of decidual CD56+ lymphocytes is influenced by secreted factors from decidual stromal cells but not macrophages in the first trimester of pregnancy.
The phenotype of decidual CD56+ lymphocytes is influenced by secreted factors from decidual stromal cells but not macrophages in the first trimester of pregnancy.
Journal of reproductive immunology (2020-01-27)
Laura B James-Allan, Rebecca J Buckley, Guy S Whitley, Judith E Cartwright
ABSTRACT
During the first trimester of pregnancy the decidua is comprised of decidual stromal cells (DSC), invading fetal trophoblast cells and maternal leukocytes, including decidual natural killer (dNK) cells and macrophages. dNK cells are distinct from peripheral blood NK cells and have a role in regulating trophoblast invasion and spiral artery remodelling. The unique phenotype of dNK cells results from the decidual environment in which they reside, however the interaction and influence of other cells in the decidua on dNK phenotype is unknown. We isolated first trimester DSC and decidual macrophages and investigated the effect that DSC and decidual macrophage secreted factors have on CD56+ decidual lymphocyte receptor expression and cytokine secretion (including dNK cells). We report that DSC secreted factors induce the secretion of the cytokines IL-8 and IL-6 from first trimester CD56+ cells. However, neither DSC nor decidual macrophage secreted factors changed CD56+ cell receptor expression. These results suggest that secreted factors from DSC influence CD56+ decidual lymphocytes during the first trimester of pregnancy and therefore may play a role in regulating the unique phenotype and function of dNK cells during placentation.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Large Column Holder, For columns o.d. 50 mm, ÄKTA®xpress, Cytiva 28400737