Merck
  • Fibrillin-1 microfibril deposition is dependent on fibronectin assembly.
Fibrillin-1 microfibril deposition is dependent on fibronectin assembly.
Journal of cell science (2008-07-26)
Rachel Kinsey, Matthew R Williamson, Shazia Chaudhry, Kieran T Mellody, Amanda McGovern, Seiichiro Takahashi, C Adrian Shuttleworth, Cay M Kielty
ABSTRACT
Newly deposited microfibrils strongly colocalise with fibronectin in primary fibroblasts. Microfibril formation is grossly inhibited by fibronectin depletion, but rescued by supplementation with exogenous cellular fibronectin. As integrin receptors are key determinants of fibronectin assembly, we investigated whether they also influenced microfibril deposition. Analysis of beta1-integrin-receptor-null fibroblasts, blockage of cell surface integrin receptors that regulate fibronectin assembly and disruption of Rho kinase all result in suppressed deposition of both fibronectin and microfibrils. Antibody activation of beta1 integrins in fibronectin-depleted cultures is insufficient to rescue microfibril assembly. In fibronectin(RGE/RGE) mutant mouse fibroblast cultures, which do not engage alpha5beta1 integrin, extracellular assembly of both fibronectin and microfibrils is markedly reduced. Thus, pericellular microfibril assembly is regulated by fibronectin fibrillogenesis.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid