Skip to Content
Merck
Tributylstannyl trifluoromethanesulfonate 90%

Tributylstannyl trifluoromethanesulfonate

Tributylstannyl trifluoromethanesulfonate 90%
Synonym(s):
Tributyltin triflate
Linear Formula:
CF3SO3Sn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
439.12

Products

Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing