Ammonium chloride
Synonym(s):
Ammonium chloride, Salmiac
Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade
Technique
Showing 1-2 of 2