
Marbofloxacin for peak identification

Synonym(s):
Marbofloxacin, 9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-piperazino)-7-oxo-7H-pyrido[1,2,3-ij][1,2,4]benzoxadiazine-6-carboxylic acid, Zeniquin®
Empirical Formula (Hill Notation):
C17H19FN4O4
CAS Number:
Molecular Weight:
362.36
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing