Delta Sleep Inducing Peptide
Synonym(s):
DSIP, Emideltide, Sleep inducing peptide, Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu
Empirical Formula (Hill Notation):
C35H48N10O15
CAS Number:
Molecular Weight:
848.81
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing