
Coumermycin A1

Empirical Formula (Hill Notation):
C55H59N5O20
CAS Number:
Molecular Weight:
1110.08
Products
Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing