
Calcium Lactobionate

Synonym(s):
Calcium lactobionate monohydrate, 4-O-β-D-Galactopyranosyl-D-gluconic acid hemicalcium salt monohydrate
Empirical Formula (Hill Notation):
C12H21O12 · 0.5Ca · H2O
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing