Skip to Content
Merck
Erythromycin tested according to Ph. Eur.

Erythromycin

Erythromycin tested according to Ph. Eur.
Empirical Formula (Hill Notation):
C37H67NO13
CAS Number:
Molecular Weight:
733.93
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Physical Form
Product Line
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Technique
Showing 1-3 of 3