Skip to Content
Merck
Reserpine

Reserpine

Reserpine
Synonym(s):
(3β, 16β, 17α, 18β, 20α)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester, Reserpine
Empirical Formula (Hill Notation):
C33H40N2O9
CAS Number:
Molecular Weight:
608.68

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Research Area
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing