Skip to Content
Merck
2-Keto-3-deoxy-D-gluconic acid lithium salt ≥95% (TLC)

2-Keto-3-deoxy-D-gluconic acid lithium salt

2-Keto-3-deoxy-D-gluconic acid lithium salt ≥95% (TLC)
Synonym(s):
3-Deoxy-D-erythro-2-hexulosonic acid lithium salt, KDG, Lithium 2-keto-3-deoxy-D-gluconate
Empirical Formula (Hill Notation):
C6H10O6 · xLi+
CAS Number:
Molecular Weight:
178.14 (free acid basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing