Skip to Content
Merck
2-Fluoro-1-methylpyridinium p-toluenesulfonate <5% 2-hydroxy-1-methylpyridinium p-toluenesulfonate, technical grade, ≥90%

2-Fluoro-1-methylpyridinium p-toluenesulfonate

2-Fluoro-1-methylpyridinium p-toluenesulfonate <5% 2-hydroxy-1-methylpyridinium p-toluenesulfonate, technical grade, ≥90%
Linear Formula:
C6H7FN · C7H7O3S
CAS Number:
Molecular Weight:
283.32
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Application
Markush Class
Markush Group
Reaction Type
Special Grade