Skip to Content
Merck
Modafinil ≥98% (HPLC)

Modafinil

Modafinil ≥98% (HPLC)
Synonym(s):
2-[(Diphenylmethyl)sulfinyl]­acetamide
Empirical Formula (Hill Notation):
C15H15NO2S
CAS Number:
Molecular Weight:
273.35

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing