2,3,4,6-Tetrachlorophenol solution
Empirical Formula (Hill Notation):
C6H2Cl4O
CAS Number:
Molecular Weight:
231.89
Products
Brand
Formula Weight
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing