Potassium hydrogen phthalate
Synonym(s):
Potassium hydrogen phthalate, Potassium phthalate monobasic, KHP, Phthalic acid monopotassium salt, Potassium biphthalate
Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Products
Product Category
Brand
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing