(1R,2R)-1-Amino-2-benzyloxycyclopentane
Synonym(s):
(1R)-trans-2-(Phenylmethoxy)cyclopentanamine, (1R,2R)-trans-2-Benzyloxy-cyclopentylamine
Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
Products
Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing