Skip to Content
Merck
D-(−)-Ribose suitable for cell culture, BioReagent

D-(−)-Ribose

D-(−)-Ribose suitable for cell culture, BioReagent
Synonym(s):
D(-)-Ribose
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing