Pepsin from porcine gastric mucosa
Synonym(s):
Pepsin A, Pepsin from hog stomach, Pepsin, Pepsin, Porcine Stomach Mucosa
CAS Number:
Molecular Weight:
35000
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Agency
Tissue
Quality Segment
Application
Showing 1-13 of 13
Compare
Product Number
Description
Pricing