Skip to Content
GB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
Indium tin oxide
Synonym(s):
ITO, Indium tin oxide
CAS Number:
50926-11-9
Products
Brand
Sigma-Aldrich (3)
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
544876
Indium tin oxide,
nanopowder, <50 nm particle size
Expand
View Pricing
790346
Indium tin oxide,
30 nm (SEM)
Expand
View Pricing
494682
Indium tin oxide,
−325 mesh, ≥99.99% trace metals basis
Expand
View Pricing