Skip to Content
Merck
Ammonium phosphate monobasic suitable for plant cell culture

Ammonium phosphate monobasic

Ammonium phosphate monobasic suitable for plant cell culture
Synonym(s):
mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate, Ammonium dihydrogen phosphate, Ammonium phosphate monobasic
Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique