Skip to Content
Merck
All Photos(3)

Key Documents

V900417

Sigma-Aldrich

Adenosine

Vetec, reagent grade, 98%

Synonym(s):

9-β-D-Ribofuranosyladenine, Adenine riboside, Adenine-9-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
Beilstein:
93029
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

234-236 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7-,10-/m1/s1

InChI key

OIRDTQYFTABQOQ-KQYNXXCUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Endogenous neurotransmitter at adenosine receptors. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti−inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Jansen et al.
Hepatology (Baltimore, Md.), 18(1), 146-152 (1993-07-01)
We covalently coupled 9-beta-D-arabinofuranosyladenine 5'-monophosphate (ara-AMP) to the carrier molecule lactosaminated human serum albumin using a water-soluble carbodiimide with a two-step conjugation method (pH 4.5 and pH 7.5) instead of the commonly used single-step conjugation at pH 7.5. This resulted
Deepak Mittal et al.
Cancer research, 74(14), 3652-3658 (2014-07-06)
Adenosine targeting is an attractive new approach to cancer treatment, but no clinical study has yet examined adenosine inhibition in oncology despite the safe clinical profile of adenosine A2A receptor inhibitors (A2ARi) in Parkinson disease. Metastasis is the main cause
S W Jansen et al.
Scientific reports, 5, 11525-11525 (2015-06-20)
Few studies have included subjects with the propensity to reach old age in good health, with the aim to disentangle mechanisms contributing to staying healthier for longer. The hypothalamic-pituitary-thyroid (HPT) axis maintains circulating levels of thyroid stimulating hormone (TSH) and
Dennis T L Wong et al.
Journal of the American College of Cardiology, 63(18), 1904-1912 (2014-03-25)
The goal of this study was to compare the diagnostic accuracy of combined computed tomography perfusion (CTP) + computed tomography angiography (CTA), transluminal attenuation gradient by 320-detector row computed tomography (TAG320) + CTA, and CTP + TAG320 + CTA (multidetector computed tomography-integrated protocol [MDCT-IP])
Kenneth W Mahaffey et al.
Journal of the American College of Cardiology, 63(15), 1493-1499 (2014-02-25)
This study sought to report the treatment effect of ticagrelor on myocardial infarction (MI) and the strategy for and impact of event adjudication in the PLATO (Platelet Inhibition and Patient Outcomes) trial. In PLATO, ticagrelor reduced cardiovascular death, MI, or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service